Anti-SYP Mouse Monoclonal Antibody [clone: 7H12]

Supplier: Abnova
MAB10369
10740-272EA 428.44 USD
10740-272
Anti-SYP Mouse Monoclonal Antibody [clone: 7H12]
Antibodies
Mouse monoclonal antibody raised against recombinant SYP.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: SYP
Clonality: Monoclonal
Clone: 7H12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR