Anti-TBX5 Mouse Monoclonal Antibody [clone: 7B11]

Supplier: Abnova
MAB10370
10740-710EA 428.44 USD
10740-710
Anti-TBX5 Mouse Monoclonal Antibody [clone: 7B11]
Antibodies
Mouse monoclonal antibody raised against recombinant TBX5.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: TBX5
Clonality: Monoclonal
Clone: 7B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR