Anti-WNT5A Mouse Monoclonal Antibody [clone: 6F2]

Supplier: Abnova
MAB10373
10744-682EA 428.44 USD
10744-682
Anti-WNT5A Mouse Monoclonal Antibody [clone: 6F2]
Antibodies
Mouse monoclonal antibody raised against recombinant WNT5A.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: WNT5A
Clonality: Monoclonal
Clone: 6F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR