Anti-HDAC3 Mouse Monoclonal Antibody [clone: 3A7B5]

Supplier: Abnova
MAB10374
10717-082EA 428 USD
10717-082
Anti-HDAC3 Mouse Monoclonal Antibody [clone: 3A7B5]
Antibodies
Mouse monoclonal antibody raised against partial recombinant HDAC3.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: HDAC3
Clonality: Monoclonal
Clone: 3A7B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR