Anti-3-DG-imidazolone Monoclonal Antibody [clone: JNH 27]

Supplier: Abnova
MAB1738
10683-118EA 747 USD
10683-118
Anti-3-DG-imidazolone Monoclonal Antibody [clone: JNH 27]
Antibodies
Mouse monoclonal antibody raised against 3-DG-imidazolone.

Recommended Dilutions: Immunohistochemistry: 7 ?g/ml

Type: Primary
Antigen: 3-DG-imidazolone
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: JNH 27
Epitope:
Host: IgG1
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR