Anti-FOXC2 Mouse Monoclonal Antibody [clone: A3]

Supplier: Abnova
MAB1739
10685-314EA 747 USD
10685-314
Anti-FOXC2 Mouse Monoclonal Antibody [clone: A3]
Antibodies
Mouse monoclonal antibody raised against native MFH.

Recommended Dilutions: Immunohistochemistry: 1-2 ?g/ml; Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: MFH
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: A3
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR