Anti-HMGB1 Mouse Monoclonal Antibody [clone: J2E1]

Supplier: Abnova
MAB1771
10717-620EA 329 USD
10717-620
Anti-HMGB1 Mouse Monoclonal Antibody [clone: J2E1]
Antibodies
Mouse monoclonal antibody raised against partial recombinant HMGB1.

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunohistochemistry: 1:100-1:300

Type: Primary
Antigen: HMGB1
Clonality: Monoclonal
Clone: J2E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR