Anti-LYSMD1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22099
10724-302EA 428.55 USD
10724-302
Anti-LYSMD1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant LYSMD1.

Recommended Dilutions: Western Blot: 1:250-1:500

Type: Primary
Antigen: LYSMD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR