Anti-MAP3K1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0513
10680-630EA 426.15 USD
10680-630
Anti-MAP3K1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP3K1.

Recommended Dilutions: ELISA: 1:1000; Dot Blot: 1:100-500

Type: Primary
Antigen: MAP3K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR