Anti-BNIP3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0264
10691-710EA 345.2 USD
10691-710
Anti-BNIP3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of BNIP3.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:1000-1:5000; Immunohistochemistry: Frozen sections; Immunoprecipitation: 1:50-1:200; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: BNIP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Clawed frog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR