Anti-MALT1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0270
10684-842EA 329 USD
10684-842
Anti-MALT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant MALT1.

Type: Primary
Antigen: MALT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR