Anti-MYC Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10345
10726-316EA 428.44 USD
10726-316
Anti-MYC Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of MYC-tag.

Recommended Dilutions: ELISA: 1:135000; Western Blot: 1:500-1:5000

Type: Primary
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR