Anti-KLH Polyclonal Antibody

Supplier: Abnova
PAB10347
10627-894EA 1178.46 USD
10627-894
Anti-KLH Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against KLH (Keyhole Limpet Hemocyanin).

Recommended Dilutions: ELISA: 1:400000; Western Blot: 1:40000; Immunohistochemistry: 1:20000

Type: Primary
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR