Anti-FOXK1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20992
10715-094EA 428.55 USD
10715-094
Anti-FOXK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant FOXK1.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: FOXK1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR