Anti-SVOPL Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21004
10740-132EA 428.55 USD
10740-132
Anti-SVOPL Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant SVOPL.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500

Type: Primary
Antigen: SVOPL
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR