Anti-SAMD8 Polyclonal Antibody

Supplier: Abnova
PAB10381
10731-368EA 786.15 USD
10731-368
Anti-SAMD8 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant SAMD8.

Recommended Dilutions: Western Blot: 5 to 10 ?g/ml

Type: Primary
Antigen: SAMD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR