Anti-ATF7IP Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21581
10690-330EA 428.55 USD
10690-330
Anti-ATF7IP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant ATF7IP.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500

Type: Primary
Antigen: ATF7IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR