Anti-SMPD3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10403
10738-746EA 428 USD
10738-746
Anti-SMPD3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of SMPD3.

Recommended Dilutions: Western Blot: 1 to 5 mg/ml

Type: Primary
Antigen: SMPD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR