Anti-PPP1CC Polyclonal Antibody

Supplier: Abnova
PAB10414
10729-872EA 428 USD
10729-872
Anti-PPP1CC Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PPP1CC.

Recommended Dilutions: Western Blot: 5 to 10 ?g/ml

Type: Primary
Antigen: PPP1CC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR