Anti-PPP2R1A Polyclonal Antibody

Supplier: Abnova
PAB10416
10729-954EA 428 USD
10729-954
Anti-PPP2R1A Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PPP2R1A.

Recommended Dilutions: Western Blot: 5 to 10 ?g/ml; Immunocy to chemistry 10 to 15 ?g/ml.

Type: Primary
Antigen: PPP2R1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR