Anti-PPP2R1A Polyclonal Antibody

Supplier: Abnova
PAB10417
10729-956EA 428 USD
10729-956
Anti-PPP2R1A Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against synthetic peptide of PPP2R1A.

Recommended Dilutions: Western Blot: 1 to 5 ?g; Immunocy to chemistry (1:500 to 1:1000

Type: Primary
Antigen: PPP2R1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR