Anti-TLR3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0341
10741-506EA 530.32 USD
10741-506
Anti-TLR3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Tlr3.

Type: Primary
Antigen: TLR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR