Anti-IL20RA Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0346
10719-136EA 428.44 USD
10719-136
Anti-IL20RA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of IL20RA.

Type: Primary
Antigen: IL20RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR