Anti-Histone H3 (K9me2) Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0653
10684-906EA 531 USD
10684-906
Anti-Histone H3 (K9me2) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of histone H3 (dimethylated lysine 9).

Type: Primary
Antigen: Histone H3 (K9me2)
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR