Anti-KIAA0907 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20449
10722-266EA 428.55 USD
10722-266
Anti-KIAA0907 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant KIAA0907.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: KIAA0907
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR