Anti-RTN4R Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0358
10734-440EA 530.32 USD
10734-440
Anti-RTN4R Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of RTN4R.

Type: Primary
Antigen: RTN4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR