Anti-IgG Goat Monoclonal Antibody (Texas Red®)

Supplier: Abnova
PAB10440
10637-406EA 346 USD
10637-406
Anti-IgG Goat Monoclonal Antibody (Texas Red®)
Antibodies
Goat anti-bovine IgG recognizes bovine IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host: Goat
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR