Anti-GPATCH2L Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21050
10703-608EA 428.55 USD
10703-608
Anti-GPATCH2L Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C14orf118.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: C14orf118
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR