Anti-PROC Sheep Polyclonal Antibody

Supplier: Abnova
PAB0687
10628-550EA 329 USD
10628-550
Anti-PROC Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against native Proc.

Recommended Dilutions: Western Blot: 25 ?g/ml

Type: Primary
Antigen: PROC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR