Anti-NTF3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0705
10727-740EA 329 USD
10727-740
Anti-NTF3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of NTF3.

Recommended Dilutions: Immunohistochemistry: 1:500-1:2000; Western Blot: 1:500-1:2000 Inhibition assay (1:10-50 in vitro 5-10 ?l/g body weight in vivo)

Type: Primary
Antigen: NTF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR