Anti-FAT1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21615
10720-170EA 428.55 USD
10720-170
Anti-FAT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant FAT1.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: FAT1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR