Anti-IgG Goat Monoclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Abnova
PAB10478
10632-350EA 346 USD
10632-350
Anti-IgG Goat Monoclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Goat anti-cat IgG recognizes cat IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Alkaline Phosphatase.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(c)
Host: Goat
Isotype:
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR