Anti-PTH2R Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0720
10730-596EA 406.15 USD
10730-596
Anti-PTH2R Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Pth2r.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: PTH2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR