Anti-NTRK3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0735
10727-770EA 329 USD
10727-770
Anti-NTRK3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant NTRK3.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:3000

Type: Primary
Antigen: NTRK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR