Anti-ATG7 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0742
10690-406EA 329 USD
10690-406
Anti-ATG7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ATG7.

Recommended Dilutions: Immunohistochemistry: 1:200-1:3000; Immunofluorescence: 1:200-1:3000

Type: Primary
Antigen: ATG7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR