Anti-FGF2 Sheep Polyclonal Antibody

Supplier: Abnova
PAB0746
10720-256EA 329 USD
10720-256
Anti-FGF2 Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against full length recombinant FGF2.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: FGF2
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Sheep
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR