Anti-HSPB1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB2047
10681-340EA 329 USD
10681-340
Anti-HSPB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of HSPB1.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-100

Type: Primary
Antigen: HSPB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR