Anti-SNCA Sheep Polyclonal Antibody

Supplier: Abnova
PAB0750
10738-778EA 406.15 USD
10738-778
Anti-SNCA Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal antibody raised against synthetic peptide of SNCA.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2000 The optimal working dilutions must be determined by the end user.

Type: Primary
Antigen: SNCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR