Anti-IgG Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Abnova
PAB10511
10735-940EA 345.85 USD
10735-940
Anti-IgG Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Rabbit anti-chicken IgG recognizes chicken IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: F(ab')2
Host: Rabbit
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR