Anti-FHDC1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21112
10714-726EA 428.55 USD
10714-726
Anti-FHDC1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant FHDC1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: FHDC1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR