Anti-IgG Rabbit Monoclonal Antibody (Biotin)

Supplier: Abnova
PAB10542
10637-866EA 346 USD
10637-866
Anti-IgG Rabbit Monoclonal Antibody (Biotin)
Antibodies
Rabbit anti-dog IgG recognizes dog IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope: F(ab')2
Host: Rabbit
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR