Anti-IgG Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Abnova
PAB10545
10736-260EA 346 USD
10736-260
Anti-IgG Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Rabbit anti-dog IgG recognizes dog IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: F(c)
Host: Rabbit
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR