Anti-IgG Rabbit Monoclonal Antibody (Biotin)

Supplier: Abnova
PAB10548
10736-258EA 346 USD
10736-258
Anti-IgG Rabbit Monoclonal Antibody (Biotin)
Antibodies
Rabbit anti-dog IgG recognizes dog IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope: F(c)
Host: Rabbit
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR