Anti-FLG2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22207
10714-840EA 428.55 USD
10714-840
Anti-FLG2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant FLG2.

Type: Primary
Antigen: FLG2
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR