Anti-IgG Rabbit Monoclonal Antibody (Biotin)

Supplier: Abnova
PAB10560
10637-850EA 345.85 USD
10637-850
Anti-IgG Rabbit Monoclonal Antibody (Biotin)
Antibodies
Rabbit anti-donkey IgG recognizes donkey IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Donkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR