Anti-IgG Chicken Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Abnova
PAB10562
10630-234EA 345.85 USD
10630-234
Anti-IgG Chicken Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Chicken anti-goat IgG recognizes goat IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Rhodamine.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Chicken
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR