Anti-FOS Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0828
10681-124EA 329 USD
10681-124
Anti-FOS Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a synthetic peptide of FOS.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: FOS
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR