Anti-STAT4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0833
10677-298EA 329 USD
10677-298
Anti-STAT4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a synthetic peptide of Stat4.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: STAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR