Anti-SP1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0854
10681-220EA 406.15 USD
10681-220
Anti-SP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a synthetic peptide of SP1.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: SP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR