Anti-His tag Polyclonal Antibody

Supplier: Abnova
PAB0862
10681-904EA 329 USD
10681-904
Anti-His tag Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptides of His tag.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: His tag
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR